N-Methyl-4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide

Lieferant: Apollo Scientific

Synonyme: 2-[4-(N-Methylsulphamoyl)phenoxy]-6-(trifluoromethyl)pyridine

PC8879-1G
APOSPC8879-1GEA 302 EUR
APOSPC8879-1G
N-Methyl-4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
N-Methyl-4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonamide
Formel: C₁₃H₁₁F₃N₂O₃S
Molecular Weight: 332,298 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109529
CAS-Nummer: 1257535-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR