4-(2-Aminoethylthio)-6-methyl-2-(trifluoromethyl)quinoline 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC8881-1G
APOSPC8881-1GEA
303
EUR
APOSPC8881-1G
4-(2-Aminoethylthio)-6-methyl-2-(trifluoromethyl)quinoline 97%
4-(2-Aminoethylthio)-6-methyl-2-(trifluoromethyl)quinoline
Formel:
C₁₃H₁₃F₃N₂S Molecular Weight: 286,316 g/mol Schmelzpunkt: 113…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038214 CAS-Nummer: 541539-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...