3-(Difluormethoxy)benzylamin 98%

Lieferant: Apollo Scientific
Gefahr

PC8902-1G PC8902-5G PC8902-10G
APOSPC8902-1GEA 37.7 EUR
APOSPC8902-1G APOSPC8902-5G APOSPC8902-10G
3-(Difluormethoxy)benzylamin 98%
3-(Difluormethoxy)benzylamin
Formel: C₈H₉F₂NO
Lagertemperatur: Kühlschrank
MDL: MFCD00236230
CAS-Nummer: 244022-71-7
UN: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR