4-Dimethoxymethyl-2-(trifluoromethyl)pyrimidine

Lieferant: Apollo Scientific

PC8909-1G
APOSPC8909-1GEA 348 EUR
APOSPC8909-1G
4-Dimethoxymethyl-2-(trifluoromethyl)pyrimidine
4-Dimethoxymethyl-2-(trifluoromethyl)pyrimidine
Formel: C₈H₉F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05662688
CAS-Nummer: 878760-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR