2-(Trifluoromethyl)pyrimidine-4-carboxamide
Lieferant: Apollo Scientific
|
Achtung
|
PC8910-1G
APOSPC8910-1GEA
245
EUR
APOSPC8910-1G
2-(Trifluoromethyl)pyrimidine-4-carboxamide
2-(Trifluoromethyl)pyrimidine-4-carboxamide
Formel:
C₆H₄F₃N₃O Molecular Weight: 191,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05662689 CAS-Nummer: 914348-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...