Methyl-2-(trifluoromethyl)pyrimidine-4-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC8912-1G
APOSPC8912-1GEA
371
EUR
APOSPC8912-1G
Methyl-2-(trifluoromethyl)pyrimidine-4-carboxylate
Methyl-2-(trifluoromethyl)pyrimidine-4-carboxylate
Formel:
C₇H₅F₃N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD05662690 CAS-Nummer: 878745-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...