1,3-Di-boc-2-(trifluoromethylsulphonyl)guanidine 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Bis(tert-butoxycarbonyl)-2-(trifluoromethanesulphonyl)guanidine, Goodman's Reagent, 1,3-Bis(tert-butoxycarbonyl)-2-(trifluoromethylsulphonyl)guanidine, 1,3-Di-Boc-2-(trifluoromethylsulphonyl)guanidine, 1,3-Di-Boc-2-(trifluoromethanesulphonyl)guanidine

PC8913-10G PC8913-1G PC8913-5G
APOSPC8913-10GEA 203 EUR
APOSPC8913-10G APOSPC8913-1G APOSPC8913-5G
1,3-Di-boc-2-(trifluoromethylsulphonyl)guanidine 95%
1,3-Di-boc-2-(trifluoromethylsulphonyl)guanidine
Formel: C₁₂H₂₀F₃N₃O₆S
Molecular Weight: 391.36 g/mol
Schmelzpunkt: 113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02683528
CAS-Nummer: 207857-15-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR