Perfluoro-3,6-dioxaoctane-1,8-dioic acid 98%

Lieferant: Apollo Scientific
Gefahr

PC8914-5G PC8914-1G
APOSPC8914-5GEA 65.4 EUR
APOSPC8914-5G APOSPC8914-1G
Perfluoro-3,6-dioxaoctane-1,8-dioic acid 98%
Perfluoro-3,6-dioxaoctane-1,8-dioic acid
Formel: C₆H₂F₈O₆
Siedepunkt: 115 °C/0,8mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD03094537
CAS-Nummer: 55621-21-1
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR