4-Bromo-2-(trifluoromethyl)-1,3-benzoxazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-(trifluoromethyl)benzo[d]oxazole

PC8917-1G
APOSPC8917-1GEA 306 EUR
APOSPC8917-1G
4-Bromo-2-(trifluoromethyl)-1,3-benzoxazole
4-Bromo-2-(trifluoromethyl)-1,3-benzoxazole
Formel: C₈H₃BrF₃NO
Molecular Weight: 266,015 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17010236
CAS-Nummer: 217326-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR