Methyl-3-[(4-fluorobenzyl)oxy]-2-thiophenecarboxylate

Lieferant: Apollo Scientific

PC8923-1G
APOSPC8923-1GEA 128 EUR
APOSPC8923-1G
Methyl-3-[(4-fluorobenzyl)oxy]-2-thiophenecarboxylate
Methyl-3-[(4-fluorobenzyl)oxy]-2-thiophenecarboxylate
Formel: C₁₃H₁₁FO₃S
Molecular Weight: 266,29 g/mol
Schmelzpunkt: 90…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00138942
CAS-Nummer: 338417-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR