{4-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanol

Lieferant: Apollo Scientific

Synonyme: 5-(Hydroxymethyl)-4-methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole

PC8925-500MG
APOSPC8925-500MGEA 268 EUR
APOSPC8925-500MG
{4-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanol
{4-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanol
Formel: C₁₂H₁₀F₃NOS
Molecular Weight: 273,28 g/mol
Schmelzpunkt: 50…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08276952
CAS-Nummer: 668477-94-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR