1,1,1-Trifluoro-3-{[3-(trifluoromethyl)benzyl]amino}-2-propanol

Lieferant: Apollo Scientific

PC8926-500MG
APOSPC8926-500MGEA 400 EUR
APOSPC8926-500MG
1,1,1-Trifluoro-3-{[3-(trifluoromethyl)benzyl]amino}-2-propanol
1,1,1-Trifluoro-3-{[3-(trifluoromethyl)benzyl]amino}-2-propanol
Formel: C₁₁H₁₁F₆NO
Schmelzpunkt: 65…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01871640
CAS-Nummer: 478068-12-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR