2,3,4-Trifluorzimtsäure 98%
Lieferant: Apollo Scientific
Synonyme:
2,3,4-Trifluorcinnamsäure, 3-(2,3,4-Trifluorphenyl)propensäure
PC8934-1G
APOSPC8934-1GEA
58
EUR
APOSPC8934-1G
2,3,4-Trifluorzimtsäure 98%
2,3,4-Trifluorzimtsäure
Formel:
C₉H₅F₃O₂ Molecular Weight: 202.13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00083552 CAS-Nummer: 207742-85-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...