4-Fluoro-1H-indazole-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC8937-1G
APOSPC8937-1GEA 353 EUR
APOSPC8937-1G
4-Fluoro-1H-indazole-5-carboxylic acid
4-Fluoro-1H-indazole-5-carboxylic acid
Formel: C₈H₅FN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11007930
CAS-Nummer: 1041481-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR