4-(Difluormethoxy)-3-hydroxybenzaldehyd 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Difluoromethoxy)-3-hydroxybenzaldehyde
PC8950-25G
APOSPC8950-25GEA
179
EUR
APOSPC8950-25G
4-(Difluormethoxy)-3-hydroxybenzaldehyd 97%
4-Difluoromethoxy-3-hydroxybenzaldehyde
Formel:
C₈H₆F₂O₃ Molecular Weight: 188.1283 g/mol Schmelzpunkt: 83…85 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04406687 CAS-Nummer: 151103-08-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...