2,3-Difluor-4-hydroxybenzaldehyd 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC8952-1G
PC8952-5G
PC8952-10G
APOSPC8952-1GEA
40.6
EUR
APOSPC8952-1G
APOSPC8952-5G
APOSPC8952-10G
2,3-Difluor-4-hydroxybenzaldehyd 97%
2,3-Difluoro-4-hydroxybenzaldehyde
Formel:
C₇H₄F₂O₂ Schmelzpunkt: 140…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08235194 CAS-Nummer: 676500-39-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...