[4-(Trifluoromethyl)pyridin-3-yl]methylamine
Lieferant: Apollo Scientific
Synonyme:
3-(Aminomethyl)-4-(trifluoromethyl)pyridine
PC8959-1G
APOSPC8959-1GEA
371
EUR
APOSPC8959-1G
[4-(Trifluoromethyl)pyridin-3-yl]methylamine
[4-(Trifluoromethyl)pyridin-3-yl]methylamine
Formel:
C₇H₇F₃N₂ Molecular Weight: 176,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06201889 CAS-Nummer: 771580-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...