2-Chlor-3-fluorisonicotinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-3-fluoropyridine-4-carboxylic acid

PC8963-1G
APOSPC8963-1GEA 21.9 EUR
APOSPC8963-1G
2-Chlor-3-fluorisonicotinsäure
2-Chlor-3-fluorisonicotinsäure
Formel: C₆H₃ClFNO₂
Molecular Weight: 175,55 g/mol
Schmelzpunkt: 199 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD04038325
CAS-Nummer: 628691-93-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR