3-Iodo-2-(trifluoromethyl)pyridine
Lieferant: Apollo Scientific
|
Achtung
|
PC8980-1G
APOSPC8980-1GEA
386
EUR
APOSPC8980-1G
3-Iodo-2-(trifluoromethyl)pyridine
3-Iodo-2-(trifluoromethyl)pyridine
Formel:
C₆H₃F₃IN Schmelzpunkt: 42…43 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08234938 CAS-Nummer: 590371-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...