4-Iodo-2-(trifluoromethyl)pyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC8981-1G
PC8981-250MG
PC8981-5G
APOSPC8981-1GEA
101
EUR
APOSPC8981-1G
APOSPC8981-250MG
APOSPC8981-5G
4-Iodo-2-(trifluoromethyl)pyridine 95%
4-Iodo-2-(trifluoromethyl)pyridine
Formel:
C₆H₃F₃IN Schmelzpunkt: 24…26 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08234939 CAS-Nummer: 590371-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...