6-Chloro-2-iodo-3-(trifluoromethyl)pyridine 95%

Lieferant: Apollo Scientific
Achtung

PC8982-1G
APOSPC8982-1GEA 521 EUR
APOSPC8982-1G
6-Chloro-2-iodo-3-(trifluoromethyl)pyridine 95%
6-Chloro-2-iodo-3-(trifluoromethyl)pyridine
Formel: C₆H₂ClF₃IN
Lagertemperatur: Kühlschrank
MDL: MFCD08235251
CAS-Nummer: 518057-64-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR