2,5-Difluoro-4-nitroaniline 98%

Lieferant: Apollo Scientific

PC900013-1G
APOSPC900013-1GEA 203 EUR
APOSPC900013-1G
2,5-Difluoro-4-nitroaniline 98%
2,5-Difluoro-4-nitroaniline
Formel: C₆H₄F₂N₂O₂
Molecular Weight: 174,105 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11519059
CAS-Nummer: 1542-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR