1-Fluoro-2-isopropoxybenzene 95%

Lieferant: Apollo Scientific

PC900048-1G
APOSPC900048-1GEA 377 EUR
APOSPC900048-1G
1-Fluoro-2-isopropoxybenzene 95%
1-Fluoro-2-isopropoxybenzene
Formel: C₉H₁₁FO
Molecular Weight: 154,181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18447593
CAS-Nummer: 1160293-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR