3-Ethoxy-4-fluorobenzaldehyde 96%
Lieferant: Apollo Scientific
PC900173-1G
APOSPC900173-1GEA
88.6
EUR
APOSPC900173-1G
3-Ethoxy-4-fluorobenzaldehyde 96%
3-Ethoxy-4-fluorobenzaldehyde
Formel:
C₉H₉FO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12022599 CAS-Nummer: 870837-27-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...