3-(Cyclopropylmethoxy)-4-fluorobenzaldehyde 96%

Lieferant: Apollo Scientific

PC900175-1G
APOSPC900175-1GEA 632 EUR
APOSPC900175-1G
3-(Cyclopropylmethoxy)-4-fluorobenzaldehyde 96%
3-(Cyclopropylmethoxy)-4-fluorobenzaldehyde
Formel: C₁₁H₁₁FO₂
Molecular Weight: 194,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24675206
CAS-Nummer: 1208231-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR