2,4-Dichloro-3,5-difluorophenol 98%

Lieferant: Apollo Scientific

PC900180-1G
APOSPC900180-1GEA 435 EUR
APOSPC900180-1G
2,4-Dichloro-3,5-difluorophenol 98%
2,4-Dichloro-3,5-difluorophenol
Formel: C₆H₂Cl₂F₂O
Molecular Weight: 198,982 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23111013
CAS-Nummer: 1394969-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR