2-Fluoro-6-(pyrrolidin-1-yl)pyridine 97%

Lieferant: Apollo Scientific

PC900195-1G
APOSPC900195-1GEA 574 EUR
APOSPC900195-1G
2-Fluoro-6-(pyrrolidin-1-yl)pyridine 97%
2-Fluoro-6-(pyrrolidin-1-yl)pyridine
Formel: C₉H₁₁FN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10699460
CAS-Nummer: 1000981-50-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR