Sodium difluoromethanesulphinate 95%

Lieferant: Apollo Scientific

PC900211-1G PC900211-250MG
APOSPC900211-1GEA 0 EUR
APOSPC900211-1G APOSPC900211-250MG
Sodium difluoromethanesulphinate 95%
Sodium difluoromethanesulphinate
Formel: CHF₂O₂S·Na
Lagertemperatur: Raumtemperatur
MDL: MFCD24113355
CAS-Nummer: 275818-95-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR