2-Chloro-6-fluoro-3-hydroxybenzaldehyde 96%

Lieferant: Apollo Scientific

PC900238-1G
APOSPC900238-1GEA 632 EUR
APOSPC900238-1G
2-Chloro-6-fluoro-3-hydroxybenzaldehyde 96%
2-Chloro-6-fluoro-3-hydroxybenzaldehyde
Formel: C₇H₄ClFO₂
Molecular Weight: 174,557 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26687879
CAS-Nummer: 1083103-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR