4-Fluor-3-methoxybenzylamin 95%

Lieferant: Apollo Scientific

Synonyme: 5-Aminomethyl-2-fluoroanisole, 5-(Aminomethyl)-2-fluoroanisole

PC900427-10G
APOSPC900427-10GEA 991 EUR
APOSPC900427-10G
4-Fluor-3-methoxybenzylamin 95%
4-Fluor-3-methoxybenzylamin
Formel: C₈H₁₀FNO
Molecular Weight: 155,1700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04116362
CAS-Nummer: 508177-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR