6-Fluor-2-chinolinol 98%

Lieferant: Apollo Scientific

PC900541-1G PC900541-5G
APOSPC900541-1GEA 0 EUR
APOSPC900541-1G APOSPC900541-5G
6-Fluor-2-chinolinol 98%
6-Fluor-2-chinolinol
Formel: C₉H₆FNO
Schmelzpunkt: 270…275 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08236766
CAS-Nummer: 22614-75-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR