2-Ethoxy-5-fluoropyridine 97%

Lieferant: Apollo Scientific

PC900585-1G PC900585-5G
APOSPC900585-1GEA 87 EUR
APOSPC900585-1G APOSPC900585-5G
2-Ethoxy-5-fluoropyridine 97%
2-Ethoxy-5-fluoropyridine
Formel: C₇H₈FNO
Molecular Weight: 141.143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18914532
CAS-Nummer: 858675-61-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR