2-Ethoxy-5-fluoropyridine 97%
Lieferant: Apollo Scientific
PC900585-1G
PC900585-5G
APOSPC900585-1GEA
87
EUR
APOSPC900585-1G
APOSPC900585-5G
2-Ethoxy-5-fluoropyridine 97%
2-Ethoxy-5-fluoropyridine
Formel:
C₇H₈FNO Molecular Weight: 141.143 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18914532 CAS-Nummer: 858675-61-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...