2-(Cyclopropylmethoxy)-5-fluoropyridine 95%

Lieferant: Apollo Scientific

PC900587-1G
APOSPC900587-1GEA 270 EUR
APOSPC900587-1G
2-(Cyclopropylmethoxy)-5-fluoropyridine 95%
2-(Cyclopropylmethoxy)-5-fluoropyridine
Formel: C₉H₁₀FNO
Molecular Weight: 167,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18914534
CAS-Nummer: 1305322-92-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR