4-(5-Fluoropyridin-2-yl)morpholine 98%

Lieferant: Apollo Scientific

Synonyme: 5-Fluoro-2-morpholinopyridine

PC900590-1G
APOSPC900590-1GEA 270 EUR
APOSPC900590-1G
4-(5-Fluoropyridin-2-yl)morpholine 98%
4-(5-Fluoropyridin-2-yl)morpholine
Formel: C₉H₁₁FN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD18089238
CAS-Nummer: 1287217-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR