3-Amino-4-fluorobenzaldehyde 95%

Lieferant: Apollo Scientific

PC900631-1G
APOSPC900631-1GEA 0 EUR
APOSPC900631-1G
3-Amino-4-fluorobenzaldehyde 95%
3-Amino-4-fluorobenzaldehyde
Formel: C₇H₆FNO
Molecular Weight: 139.127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09955452
CAS-Nummer: 1005507-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR