4,6-Difluorisatin 98%

Lieferant: Apollo Scientific

PC900686-25G
APOSPC900686-25GEA 97.8 EUR
APOSPC900686-25G
4,6-Difluorisatin 98%
4,6-Difluorisatin
Formel: C₈H₃F₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03618552
CAS-Nummer: 126674-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR