N-[3,5-Bis(trifluoromethyl)benzenesulphonyl]-L-methionine methyl ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl (2S)-2-({[3,5-bis(trifluoromethyl)phenyl]sulphonyl}amino)-4-(methylthio)butanoate

PC9007-100MG
APOSPC9007-100MGEA 43.5 EUR
APOSPC9007-100MG
N-[3,5-Bis(trifluoromethyl)benzenesulphonyl]-L-methionine methyl ester 97%
N-[3,5-Bis(trifluoromethyl)benzenesulphonyl]-L-methionine methyl ester
Formel: C₁₄H₁₅F₆NO₄S₂
Molecular Weight: 439,4 g/mol
Schmelzpunkt: 99…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084899
CAS-Nummer: 175202-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR