(2,4-difluoro-3-methylphenyl)boronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (2,4-Difluoro-3-methylphenyl)boronic acid, 2,4-Difluoro-3-methylphenylboronic acid

PC900770-1G
APOSPC900770-1GEA 242 EUR
APOSPC900770-1G
(2,4-difluoro-3-methylphenyl)boronic acid 98%
(2,4-difluoro-3-methylphenyl)boronic acid
Formel: C₇H₇BF₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23712890
CAS-Nummer: 1619980-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR