5-Fluoro-6-methylpyridine-2-boronic acid 95%
Lieferant: Apollo Scientific
Synonyme:
(5-Fluoro-6-methylpyridin-2-yl)boronic acid
PC900779-1G
APOSPC900779-1GEA
766
EUR
APOSPC900779-1G
5-Fluoro-6-methylpyridine-2-boronic acid 95%
5-Fluoro-6-methylpyridine-2-boronic acid
Formel:
C₆H₇BFNO₂ Molecular Weight: 154,935 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08061580 CAS-Nummer: 1208101-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...