2-Ethoxy-6-fluorphenylboronsäure 98%

Lieferant: Apollo Scientific

PC900865-25G
APOSPC900865-25GEA 149 EUR
APOSPC900865-25G
2-Ethoxy-6-fluorphenylboronsäure 98%
2-Ethoxy-6-fluorphenylboronsäure
Formel: C₈H₁₀BFO₃
Molecular Weight: 183.97 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05664322
CAS-Nummer: 957062-68-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR