1-Acetyl-3,5-bis(trifluoromethyl)-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

PC9009-5G
APOSPC9009-5GEA 71.3 EUR
APOSPC9009-5G
1-Acetyl-3,5-bis(trifluoromethyl)-1H-pyrazole
1-Acetyl-3,5-bis(trifluoromethyl)-1H-pyrazole
Formel: C₇H₄F₆N₂O
Molecular Weight: 246,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153613
CAS-Nummer: 244187-01-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR