[4-Fluoro-2-(methoxymethoxy)phenyl]boronic acid 98%

Lieferant: Apollo Scientific

PC900914-1G
APOSPC900914-1GEA 328 EUR
APOSPC900914-1G
[4-Fluoro-2-(methoxymethoxy)phenyl]boronic acid 98%
[4-Fluoro-2-(methoxymethoxy)phenyl]boronic acid
Formel: C₈H₁₀BFO₄
Molecular Weight: 199,972 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24122672
CAS-Nummer: 335254-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR