2-Fluoro-3-methoxy-5-methylphenylboronic acid 98%

Lieferant: Apollo Scientific

PC901033-1G
APOSPC901033-1GEA 364 EUR
APOSPC901033-1G
2-Fluoro-3-methoxy-5-methylphenylboronic acid 98%
2-Fluoro-3-methoxy-5-methylphenylboronic acid
Formel: C₈H₁₀BFO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD19440599
CAS-Nummer: 2096329-52-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR