2-(3-Fluorophenyl)piperidine 95%

Lieferant: Apollo Scientific

PC901105-1G
APOSPC901105-1GEA 498 EUR
APOSPC901105-1G
2-(3-Fluorophenyl)piperidine 95%
2-(3-Fluorophenyl)piperidine
Formel: C₁₁H₁₄FN
Molecular Weight: 179,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02663623
CAS-Nummer: 383128-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR