4-Cyano-3,5-difluorophenylboronic acid 95%

Lieferant: Apollo Scientific

Synonyme: (4-Cyano-3,5-difluorophenyl)boronic acid

PC901287-1G
APOSPC901287-1GEA 163 EUR
APOSPC901287-1G
4-Cyano-3,5-difluorophenylboronic acid 95%
4-Cyano-3,5-difluorophenylboronic acid
Formel: C₇H₄BF₂NO₂
Molecular Weight: 182,92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07644470
CAS-Nummer: 861710-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR