2-Fluoro-6-propoxypyridine-3-boronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (2-Fluoro-6-propoxypyridin-3-yl)boronic acid

PC901347-1G PC901347-5G
APOSPC901347-1GEA 128 EUR
APOSPC901347-1G APOSPC901347-5G
2-Fluoro-6-propoxypyridine-3-boronic acid 98%
2-Fluoro-6-propoxypyridine-3-boronic acid
Formel: C₈H₁₁BFNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18434411
CAS-Nummer: 2096338-78-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR