2-Fluoro-6-isopropoxypyridine-3-boronic acid 97%

Lieferant: Apollo Scientific

PC901349-1G
APOSPC901349-1GEA 364 EUR
APOSPC901349-1G
2-Fluoro-6-isopropoxypyridine-3-boronic acid 97%
2-Fluoro-6-isopropoxypyridine-3-boronic acid
Formel: C₈H₁₁BFNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18434413
CAS-Nummer: 1637749-71-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR