2-Fluoro-5-(methoxymethoxy)phenylboronic acid 97%
Lieferant: Apollo Scientific
Synonyme:
(2-Fluoro-5-(methoxymethoxy)phenyl)boronic acid
PC901368-1G
APOSPC901368-1GEA
250
EUR
APOSPC901368-1G
2-Fluoro-5-(methoxymethoxy)phenylboronic acid 97%
2-Fluoro-5-(methoxymethoxy)phenylboronic acid
Formel:
C₈H₁₀BFO₄ Molecular Weight: 199,972 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18837601 CAS-Nummer: 2096332-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...