2,6-Difluoro-3-methylphenylboronic acid 98%

Lieferant: Apollo Scientific

PC901371-1G
APOSPC901371-1GEA 249 EUR
APOSPC901371-1G
2,6-Difluoro-3-methylphenylboronic acid 98%
2,6-Difluoro-3-methylphenylboronic acid
Formel: C₇H₇BF₂O₂
Molecular Weight: 171,937 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09265479
CAS-Nummer: 1586045-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR